Orders LinkContactUsLinkComponent
Sticky Swabs, Electron Microscopy Sciences
Sticky swabs, 3 mm
  76499-604
 :  78324-65
Sticky Swabs, Electron Microscopy Sciences
  76499-604
Sticky Swabs, Electron Microscopy Sciences
Sticky Swabs, Electron Microscopy Sciences
Sticky swabs, 3 mm
54,06 $
Each
Dust-free sticky swabs are specially used and designed for a high degree dust-free environment.
  • ESD safe
  • Made of PVC
  • Available in three sizes: 1, 2, and 3 mm
Sticky swabs have an anti-static treatment that can safely dissipate electrostatic charges. They are applicable for manufacturing and assembling static-sensitive products.
 : Kit contains 25 of each size.
  • Description:
    Sticky swabs, 3 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.