Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CYBIO TIPBOX 192 60UL COLUMNS CS10
Catalog # CA76469-750
Supplier:
Analytik Jena CA
Expand image
CYBIO TIPBOX 192 60UL COLUMNS CS10
Catalog # CA76469-750
Supplier:
Analytik Jena CA
Supplier Number:
OL000825575N
Related Information
Documentation
Product Literature
Analytik Jena CyBio® FeliX_CA
Documentation
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH