Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TIPBOX 96 250UL-PCR-CERT PRE-STER CS10
Catalog # CA76469-744
Supplier:
Analytik Jena CA
Expand image
TIPBOX 96 250UL-PCR-CERT PRE-STER CS10
Catalog # CA76469-744
Supplier:
Analytik Jena CA
Supplier Number:
OL381125637S
Related Information
Documentation
Product Literature
Analytik Jena CyBio® FeliX_CA
Documentation
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH