Accessories for CyBio® FeliX Automated Pipette Robots, Analytik Jena US
Catalog # CA76469-620
Supplier: Analytik Jena CA
Specifications
- Description:Cybio Felix head R 96 60 µl
Specifications
About this item
Accessories for the CyBio FeliX automation platform including upgrades, adaptors, and support mechanisms.
- Shakers, Thermoshaker, and Heater provide mixing and temperature control of samples
- Labware adaptors are available to fit plates and reservoirs with different heights
- General accessories available for tip transfer, and tip waste to create a complete liquid handling solution
Frequently Bought Together