Order Entry
Canada
Orders LinkContactUsLinkComponent
Sera- Separa Columns, Caplugs

Sera- Separa Columns, Caplugs

Sera-Separa columns, 2.625", PE, 5 ml
Catalog # 76498-810
Supplier:   Caplugs
Supplier #:  208-3193-020
Sera- Separa Columns, Caplugs
Catalog # 76498-810
Supplier:  Caplugs
Sera- Separa Columns, Caplugs
Sera- Separa Columns, Caplugs

Sera-Separa columns, 2.625", PE, 5 ml

Price
$978.82
Log in for availability
Carton of 1,000
Product Details & Documents

Sera-Separa columns are ideal for use in removing fibrin and macro particles from serum to prevent line clogging.

  • Graduated barrel tells exactly how much liquid has been filtered
  • Wide-body style prevents sample probes from hitting side walls

Users insert the 16 mm flange into the evacuated blood tube and push downward.

Specifications
  • Capacity:
    5 ml
  • Description:
    Sera-Separa columns, 2.625"
  • Column hardware:
    PE
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.