Sera- Separa Columns, Caplugs
Sera-Separa columns, 4.25", PE, 9 ml
Catalog # 76498-808
Supplier: Caplugs
Supplier #: 208-3084-020
Jump to:
Product Details & Documents
Sera-Separa columns are ideal for use in removing fibrin and macro particles from serum to prevent line clogging.
- Graduated barrel tells exactly how much liquid has been filtered
- Wide-body style prevents sample probes from hitting side walls
Users insert the 16 mm flange into the evacuated blood tube and push downward.
Specifications
- Capacity:9 ml
- Description:Sera-Separa columns, 4.25"
- Column hardware:PE



