Autopsy Head Drape and Framework
10 head drapes, without framework
Catalog # 76498-428
Jump to:
Discontinued
Product Details & Documents
Helps contain infectious and contaminated materials during autopsy procedures. Easily assembled with a wide variety of head, torso and/or appendage dissections.
- Autopsy head drape kit includes framework and four polyethylene head drapes (0.03 mm thick, non-autoclavable)
- Easily assembled with a wide variety of head, torso and/or appendage dissections
Specifications
- Description:10 head drapes, without framework