Order Entry
Canada
Orders LinkContactUsLinkComponent
Autopsy Head Drape and Framework

Autopsy Head Drape and Framework

Autopsy head drape and framework, Polyethylene, 26×20×4.5"
Catalog # 76498-426
Supplier:   Mopec
Supplier #:  BE010
Autopsy Head Drape and Framework
Catalog # 76498-426
Supplier:  Mopec
Autopsy Head Drape and Framework
Autopsy Head Drape and Framework

Autopsy head drape and framework, Polyethylene, 26×20×4.5"

Price
$915.92
Log in for availability
Each
Product Details & Documents

Helps contain infectious and contaminated materials during autopsy procedures. Easily assembled with a wide variety of head, torso and/or appendage dissections.

  • Autopsy head drape kit includes framework and four polyethylene head drapes (0.03 mm thick, non-autoclavable)
  • Easily assembled with a wide variety of head, torso and/or appendage dissections
Specifications
  • Description:
    Autopsy head drape and framework
  • Dimensions:
    26×20×4.5"
  • Material:
    Polyethylene