Order Entry
Canada
Orders LinkContactUsLinkComponent
Dissecting Boards with Optional Legs, Mortech®

Dissecting Boards with Optional Legs, Mortech®

Dissecting board, with 3" legs, Dark blue, 16×12"
Catalog # 76498-328
Supplier:   Mopec
Supplier #:  BC068
Dissecting Boards with Optional Legs, Mortech®
Catalog # 76498-328
Supplier:  Mopec
Dissecting  Boards with Optional Legs, Mortech®
Dissecting Boards with Optional Legs, Mortech®

Dissecting board, with 3" legs, Dark blue, 16×12"

Price
$589.91
Log in for availability
Each
Product Details & Documents

Easy-to-clean dissection board with optional removable legs offer a more ergonomic multi-user work surface.

  • Polyethylene construction
  • Custom sizes available upon request

Cutting boards have safe rounded edges and removable legs or non-skid feet. Dark blue color is available for specimen photography.

Specifications
  • Color:
    Dark blue
  • Description:
    Dissecting board, with 3" legs
  • L×W:
    16×12"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.