Order Entry
Canada
Orders LinkContactUsLinkComponent
Dissecting Boards with Optional Legs, Mortech®

Dissecting Boards with Optional Legs, Mortech®

Dissecting board, with 3" legs, Dark blue, 12×11"
Catalog # 76498-324
Supplier:   Mopec
Supplier #:  BC066
Dissecting Boards with Optional Legs, Mortech®
Catalog # 76498-324
Supplier:  Mopec
Dissecting  Boards with Optional Legs, Mortech®
Dissecting Boards with Optional Legs, Mortech®

Dissecting board, with 3" legs, Dark blue, 12×11"

Price
$471.64
Log in for availability
Each
Product Details & Documents

Easy-to-clean dissection board with optional removable legs offer a more ergonomic multi-user work surface.

  • Polyethylene construction
  • Custom sizes available upon request

Cutting boards have safe rounded edges and removable legs or non-skid feet. Dark blue color is available for specimen photography.

Specifications
  • Color:
    Dark blue
  • Description:
    Dissecting board, with 3" legs
  • L×W:
    12×11"