Storage Containers with Optional Lids, Mortech®
—, Storage container lid, 2 qt. container
Catalog # 76498-186
Supplier: Mopec
Supplier #: BA022
Jump to:
Product Details & Documents
Smooth stainless steel finish for easy cleaning.
- All stainless steel, seamless construction
- Smooth finish for easy cleaning
- No pour spout
Specifications
- Capacity:—
- Description:Storage container lid
- For use with:2 qt. container
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



