$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
CARTRIDGE 10IN 1U PP BF/226 BAYONT SILCN
76495-542
:
PARKER HANNIFIN TE
CARTRIDGE 10IN 1U PP BF/226 BAYONT SILCN
76495-542
:
PARKER HANNIFIN TE
:
ZCPH1-1.0C
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Disposable Petri Dishes
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International