INSTALLATION IQ OQ AND FAMILIARIZATION
Catalog # 76493-312
Supplier: HITACHI HIGH-TECH AMERICA, INC.
INSTALLATION IQ OQ AND FAMILIARIZATION
Catalog # 76493-312
Supplier: HITACHI HIGH-TECH AMERICA, INC.
Supplier Number: IQ/OQ-2
Frequently Bought Together
VWR® Self-Seal Sterilization Pouches
Supplier: VWR International
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
Single-Use Needles, BD Medical
Supplier: BD
Sodium thiosulfate 0.100 N (N/10) in aqueous solution
Supplier: Ricca Chemical
Acetone ≥99.5% (by GC, corrected for water content), GR ACS, Supelco®
Supplier: MilliporeSigma
Corning® Fetal Bovine Serum, Regular, CA sourced, Corning
Supplier: Corning
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
PREempt® Concentrate Disinfectants, Contec
Supplier: Contec
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Bacto™ Brain Heart Infusion, BD Diagnostics
Supplier: BD
First Aid Central Hot and Cold Compresses, Acme United
Supplier: ACME UNITED LIMITED CA
Human Recombinant GDNF (from <i>E. coli</i>)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Sol-M® Hypodermic Needles
Supplier: SOL-MILLENNIUM CANADA, INC. CA