Test Tubes, Caplugs
19.0 ml, PS, Natural, 16×125 mm, Round bottom
Catalog # 76493-652
Supplier: Caplugs
: 214-2065-01N
Jump to:
Product Details & Documents
Test tubes provide clarity and can be used for mild acidic or alkali solutions, alcohol and salt, with exceptions of oxidizing acids and high concentrate aclohols.
- Provided with or without colors
- Tubes are bulk packed, non-sterile
- Polystyrene tubes withstand up to 8200×g
- Polypropylene tubes withstand up to 10000×g
- 12×75 mm tubes are available with internally ribs for use in antibody coating and/or protein adsorptions, six slanted ribs increase useful surface area
The polypropylene tubes are chemically resistant and are appropriate for use with organic solvents and stronger acids, alkalis, alcohols, and chlorinated and aromatic hydrocarbons.
Specifications
- Capacity:19.0 ml
- Color:Natural
- Material:PS
- Diameter ×H:16×125 mm
- Bottom style:Round bottom



