InocuLoop Inoculating Loops, Caplugs
Loop and spreader, 1 µl, PS, Light blue
Catalog # 76492-638
Supplier: Caplugs
Supplier #: 333-5002-B1I
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Inoculoop disposable inoculating loops have patented four-sided spreaders, enabling the inoculation of a petri dish with one, not multiple, disposable loops. Make bacterial inoculation and quantitative procedures faster and more efficient.
- Made of flexible plastic for easy spreading
- Can also be used as a picker for isolating colonies
- Pre-sterilized Inoculoops are gamma-irritated
Suitable in anaerobic chambers, in the field and even in satellite metal loops.
Specifications
- Capacity:1 µl
- Color:Light blue
- Description:Loop and spreader
- Material:PS