Orders LinkContactUsLinkComponent
Dissection Scissors
Dissection Scissors
Mayo scissors, 5.5", Stainless steel, Sharp/sharp
Catalog # 470334-870
Supplier:   AFTAB AHMED SE
Supplier #:  10-916
Dissection Scissors
Catalog # 470334-870
Supplier:  AFTAB AHMED SE
Dissection Scissors
Dissection Scissors
Mayo scissors, 5.5", Stainless steel, Sharp/sharp
Price
$8.35
Log in for availability
Each Box of 12
Product Details & Documents
Basic dissection for students.
  • Stainless steel
Basic dissection scissors are strong, durable, and have shorter handles for good control. A good option for inexperienced students, these have a slightly blunted pointed blade set to also minimize the chance for accidental pokes.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
  • Description:
    Mayo scissors
  • Length:
    5.5"
  • Material:
    Stainless steel
  • Blade type:
    Sharp/sharp