Dissection Scissors
Economy student scissors, 4.5", Stainless steel, Sharp/sharp
Catalog # 470334-864
Supplier: AFTAB AHMED SE
Supplier #: 14-0525
Jump to:
Product Details & Documents
Basic dissection for students.
- Stainless steel
Basic dissection scissors are strong, durable, and have shorter handles for good control. A good option for inexperienced students, these have a slightly blunted pointed blade set to also minimize the chance for accidental pokes.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
- Description:Economy student scissors
- Length:4.5"
- Material:Stainless steel
- Blade type:Sharp/sharp