Orders LinkContactUsLinkComponent
Dissection Scissors
Dissection Scissors
Fine tip, 4.5", Stainless steel, Sharp/sharp
Catalog # 470334-862
Supplier:   AFTAB AHMED SE
Supplier #:  14-0980
Dissection Scissors
Catalog # 470334-862
Supplier:  AFTAB AHMED SE
Dissection Scissors
Dissection Scissors
Fine tip, 4.5", Stainless steel, Sharp/sharp
Price
$6.35
Log in for availability
Each Box of 12
Product Details & Documents
Basic dissection for students.
  • Stainless steel
Basic dissection scissors are strong, durable, and have shorter handles for good control. A good option for inexperienced students, these have a slightly blunted pointed blade set to also minimize the chance for accidental pokes.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
  • Description:
    Fine tip
  • Length:
    4.5"
  • Material:
    Stainless steel
  • Blade type:
    Sharp/sharp