Dissection Needles
Straight teasing needle, Stainless steel, 5.5", Sharp
Catalog # 470334-848
Supplier #: 14-1033
Jump to:
Discontinued
Product Details & Documents
Sharp point needles.
- Fine point teasing needles
- Straight or angled tips
Very sharp points on these needles make tissue separation fast and easy. Used mostly to break apart muscle tissues.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
- Description:Straight teasing needle
- Length:5.5"
- Material:Stainless steel
- Tip style:Sharp