Bone Shears
Small bone cutting shears, 5", Stainless steel
Catalog # 470334-810
Supplier: AFTAB AHMED SE
Supplier #: 14-1110
Jump to:
Product Details & Documents
Easily snip through bones.
- Stainless steel
- Flush cutting face
- Short jaws for additional force
Used to easily cut through tough tissue or bone during dissection. These bone cutting shears combine longer grips and shorter heads to make cuts easier. The cutter has a beveled edge on one side for easy flush cutting.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
- Description:Small bone cutting shears
- Length:5"
- Material:Stainless steel