Orders LinkContactUsLinkComponent
Watchmaker Forceps
Watchmaker Forceps
Stainless steel, 4.25", Straight
Catalog # 470334-804
Supplier:   AFTAB AHMED SE
Supplier #:  14-0505
Watchmaker Forceps
Catalog # 470334-804
Supplier:  AFTAB AHMED SE
Watchmaker Forceps
Watchmaker Forceps
Stainless steel, 4.25", Straight
Price
$9.00
Log in for availability
Each Box of 12
Product Details & Documents
Ultra-fine point forceps.
  • Stainless steel
Extremely fine tips and heavier handles allow for the manipulation of very fine tissue samples with a secure hold.
These forceps have a smooth finish inside the jaws, to ensure a minimum of tissue damage.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
  • Length:
    4.25"
  • Material:
    Stainless steel
  • Tip style:
    Straight