MXP Data Loggers, Cryopak Verification Technologies
Data logger, MXP single use
Catalog # 76490-420
Supplier: Cryopak Verification Technologies
Fournisseur #: MXP-S
MXP Data Loggers, Cryopak Verification Technologies
Catalog # 76490-420
Supplier: Cryopak Verification Technologies
Jump to:
Product Details & Documents
The MXP data logger offers flexibility and longevity with a light sensor to detect and log if package is opened.
- Long shelf life
- Event logging feature
- Internal digital temperature sensor
With its USB connectivity, the MXP data logger provides detailed reports in both PDF and Excel format, and is user friendly for both Windows and smartphone app use.
Spécifications
- Description:Data logger, MXP single use