J.T.Baker® Plate 96
PP plates, U-bottom, 0,3 ml, Transparent, -
Catalog # 43001-0600
Supplier: Ritter, part of Avantor
: 43001-06001
Jump to:
Product Details & Documents
J.T.Baker® Plate 96 round well plates made from PP are ideal for ELISA applications.
- Alphanumeric coding assists with identification of samples
- Plate shape significantly reduces sample carryover
- Standardised dimensions (128×85 mm) ensure compatibility with most common laboratory machines
Documents
Specifications
- Well volume:0,3 ml
- Color:Transparent
- Description:PP plates, U-bottom
- Sterile:-
- Cat. No.:43001-0600
- Environmental certification:ISO 50001 (Certificate Registration No.) 10000406726-MSC-RvA-DEU
- Environmentally Preferable:Low Manufacturing Impact
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



