Plastic Beakers
250 ml, 10 ml
Catalog # 470332-662
Supplier: United Scientific Supplies
Supplier #: BS250-PK/6
Jump to:
Product Details & Documents
Plastic beakers for classroom activites.
- Raised graduations
- Easy pour spout
These plastic beakers are perfect additions to your classroom. The graduations are easy to read.
Specifications
- Graduation Interval:10 ml
- Capacity:250 ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



