Solid Balls for Physics Experiments
Solid Brass Ball, 25 mm, 1"
Catalog # 470332-644
Supplier: United Scientific Supplies
Supplier #: PNBB25-S
Jump to:
Product Details & Documents
Drilled balls are used for pendulum bobs or impact experiments, while our other balls are suitable for collision apparatus and density/volume experiments.
Caution: WARNING: CHOKING HAZARD -Item contains a small ball. Not for children under 3 years.
Specifications
- Description:Solid Brass Ball, 25 mm
- Diameter:1"
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



