Dextrose, anhydrous 97.5-102.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
CAJT1920-01
:
- Pack type:Poly bottle
- Size:500 g
- Cat. no.:CAJT1920-01
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Identification C: Water (H₂O):≤1.0 %
- USP - Assay (Anhydrous basis):97.5 - 102.0 %
- USP - Related substances: Maltose and isomaltose:≤0.4 %
- USP - Related substances: Maltotriose:≤0.2 %
- USP - Related substances: Fructose:≤0.15 %
- USP - Related substances: Unspecified impurities, each:≤0.10 %
- USP - Related substances: Total Impurities:≤0.5 %
- USP - Color and Clarity of Solution:Passes Test
- USP - Dextrin:Passes Test
- USP - Soluble Starch, Sulfites:≤15 ppm
- USP - Conductivity (uS cm-1):≤20 µS/cm
- USP - Endotoxin Concentration (EU/g):≤2.5
- EP/BP - Identification A (Specific Rotation (+)):52.5 - 53.3 Degree
- EP/BP - Identification B:Passes Test
- EP/BP - Identification E:Passes Test
- EP/BP - Assay (C₆H₁₂O₆) (anhydrous substance):97.5 - 102.0 %
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Conductivity, uS cm-1:≤20
- EP/BP - Related Substances: sum of impurities A and B:≤0.4 %
- EP/BP - Related Substances: impurity C:≤0.2 %
- EP/BP - Related Substances: impurity D:≤0.15 %
- EP/BP - Related Substances: unspecified impurities, each:≤0.10 %
- EP/BP - Related Substances: total impurities:≤0.5 %
- EP/BP - Dextrins:Passes Test
- EP/BP - Soluble starch, sulfite:≤15 ppm
- EP/BP - Water (H₂O):≤1.0 %
- JP - Identification 1:Passes Test
- JP - Identification 2:Passes Test
- JP - Water (H₂O):≤1.0 %
- JP - Assay (Anhydrous basis):97.5 - 102.0 %
- JP - Maltose and isomaltose:≤0.4 %
- JP - Maltotriose:≤0.2 %
- JP - Fructose:≤0.15 %
- JP - Unspecified Impurities, each:≤0.10 %
- JP - Total Impurities:≤0.5 %
- JP - Color and Clarity of Solution:Passes Test
- JP - Soluble Starch, Sulfites:≤15 ppm
- JP - Conductivity (uS cm-1):≤20 µS/cm
- JP - Dextrin:Passes Test
- JP - Heavy Metals (as Pb):≤4 ppm
:
:14,04459
:146 °C
:1.54 g/cm³ (20 °C)
:Management of Change (MOC) category = R
:180.16 g/mol
:Ambient
:200-075-1
Frequently Bought Together