Orders LinkContactUsLinkComponent
Polyethylene Dropping Bottles
Polyethylene Dropping Bottles
473 ml (16 oz.), LDPE
Catalog # 470332-528
Supplier:   Dynalon
Supplier #:  605065-0160
Polyethylene Dropping Bottles
Catalog # 470332-528
Supplier:  Dynalon
Polyethylene Dropping Bottles
Polyethylene Dropping Bottles
473 ml (16 oz.), LDPE
Price
$16.27
Log in for availability
Each Case of 24
Product Details & Documents
Economy LDPE dropper bottles.
  • Translucent LDPE dropper bottles
  • Sturdy white dropping cap
  • Flip up spout
Dropper bottles with hinged dispenser caps for dispensing indicator solutions, dyes, and more.
Specifications
  • Capacity:
    473 ml (16 oz.)
  • Material:
    LDPE