Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Certipur® Buffer Solution Sachet Sets, MilliporeSigma

Certipur® buffer solution, pH 4.00, citric acid, sodium hydroxide, hydrogen chloride, Citrate, Sachet, 4.00
Catalog # EM1.99064.0001
Supplier:   MilliporeSigma
 :  1990640001
Certipur® Buffer Solution Sachet Sets, MilliporeSigma
Catalog # EM1.99064.0001
Supplier:  MilliporeSigma
CAS Number:  
Certipur® Buffer Solution Sachet Sets, MilliporeSigma
Certipur® Buffer Solution Sachet Sets, MilliporeSigma

Certipur® buffer solution, pH 4.00, citric acid, sodium hydroxide, hydrogen chloride, Citrate, Sachet, 4.00

CAS Number:  
Price
$359.62
Log in for availability
Each (900ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

30 ml buffers sachets - ideal for the calibration of pH meters.

  • Practical 30 mL sachets – one use per sachet – no risk of contamination
  • Solutions are always fresh and ready to use
  • Batch traceable to NIST
  • Reference temperature: 25°C
  • Precision: ±0.015 pH units max.
Specifications

Specifications

  • Pack type:
    Sachet
  • Description:
    Certipur® buffer solution, pH 4.00, citric acid, sodium hydroxide, hydrogen chloride
  • pH buffer type:
    Citrate
  • pH:
    4.00

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.