Orders LinkContactUsLinkComponent
Bedside Carts, TrippNT
Bedside Carts, TrippNT
White bedside cart body with black drawer, 14⅜", 27¾", 18⅛", Hinged
Catalog # 76471-898
Supplier:   TrippNT
Supplier #:  53407
Bedside Carts, TrippNT
Catalog # 76471-898
Supplier:  TrippNT
Bedside Carts, TrippNT
Bedside Carts, TrippNT
White bedside cart body with black drawer, 14⅜", 27¾", 18⅛", Hinged
Price
$1,550.35
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
TrippNT's Bedside Cart offers total locking mobile storage.
  • 2 locking casters, 2 swivel casters, all locking drawer and bulk storage area
  • Built-in recessed tray in the top of the cart
  • Corrosion-resistant, non-metal HDPE construction
  • Designed to secures personal effects during transit and stationary use
  • 100% total replacement lifetime guarantee/made in USA
TrippNT's Bedside Cart is used to protect and store patient’s belongings. The top of the cart is inset to create a tray to secures items such as: phones, drinks, and other possessions that need to be frequently accessed. The bulk storage area and single drawer lock with a single key to secures items such as backpacks, laptops, coats, and other personal effects allowing for easy transportation. 2 of the casters are locking for stationary use as needed.
Ordering information: Available in 13 color variants.
Delivery information: Ships fully assembled.
Specifications
  • Depth:
    18⅛"
  • Width:
    14⅜"
  • Description:
    White bedside cart body with black drawer
  • Height:
    27¾"
  • Door type:
    Hinged