Order Entry
Canada
Orders LinkContactUsLinkComponent
Replacement Tips, Excelta Corp®

Replacement Tips, Excelta Corp®

Replacement tips for 159F-RT, Carbon fiber tips, 2.03 mm (0.08"), 0.05" (1.3 mm), Straight
Catalog # 76471-538
Supplier:   Excelta
Supplier #:  159F-RTX
Replacement Tips, Excelta Corp®
Catalog # 76471-538
Supplier:  Excelta
Replacement Tips, Excelta Corp®
Replacement Tips, Excelta Corp®

Replacement tips for 159F-RT, Carbon fiber tips, 2.03 mm (0.08"), 0.05" (1.3 mm), Straight

Price
$16.55
Log in for availability
Each
Product Details & Documents

High-quality forcep tips.

  • High rigidity, excellent tensile and flexural strength.

These soft replacement tips are made of a Carbofib™ material which is a polymer based material with 30% carbon fibers added. The working temperature is 249 °F. Carbofib™ has very high rigidity, excellent tensile and flexural strength, and fatigue and creep resistance. The tips are conductive (102 ohms/sq). Each includes 4 screws and 2 tips.

Specifications
  • Description:
    Replacement tips for 159F-RT
  • Material:
    Carbon fiber tips
  • Curvature:
    Straight
  • Tip width:
    0.05" (1.3 mm)
  • Tip length:
    2.03 mm (0.08")