Order Entry
Canada
Orders LinkContactUsLinkComponent
Cutters, Carbide Insert, Excelta | Avantor

Cutters, Carbide Insert, Excelta

Cutters, taper head, laser flush cut, carbide inserts, molded handle, Carbon steel, 118.75 mm (4.75"), 14.25°
Catalog # 76471-382
Supplier:   Excelta
Fournisseur #:  C7245X-GM
Cutters, Carbide Insert, Excelta
Catalog # 76471-382
Supplier:  Excelta
Cutters, Carbide Insert, Excelta
Cutters, Carbide Insert, Excelta

Cutters, taper head, laser flush cut, carbide inserts, molded handle, Carbon steel, 118.75 mm (4.75"), 14.25°

Price
$437.06
Log in for availability
Each
Product Details & Documents

This compound action cutter is ideal for cutting small hard wire.

  • Carbide insert cutting edges for tough hard wire
  • Compound-Action means less pressure on hands
  • Dual leaf springs for ease of use
  • Stainless steel shank for corrosion resistants

These multi-purpose medium round hard wire cutters feature stainless steel construction for corrosion resistance and carbide inserts for maximum blade life. Hard wire cutting capacity is 0.010 to 0.020". The grips are soft cushioned foam and have a surface resistivity of 10¹⁰ ohms/sq. Interestingly, the 149AT is a compound-action cutter, which requires much less force to operate.

Specifications
  • Description:
    Cutters, taper head, laser flush cut, carbide inserts, molded handle
  • Material:
    Carbon steel
  • Curvature:
    14.25°
  • Overall length:
    118.75 mm (4.75")
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.