Order Entry
Canada
Orders LinkContactUsLinkComponent
SmartWall® Uprights

SmartWall® Uprights

Upright, Grey, Metroseal epoxy, Steel, 1⅛", 1⅞", 84½"
Catalog # 76463-170
Supplier:   Metropolitan Wire
Supplier #:  SWU85K4
SmartWall® Uprights
Catalog # 76463-170
Supplier:  Metropolitan Wire
SmartWall® Uprights
SmartWall® Uprights

Upright, Grey, Metroseal epoxy, Steel, 1⅛", 1⅞", 84½"

Price
$474.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Create a unique storage and workstation system for underutilized wall space with SmartWall® Uprights.

  • Built-in slots allow shelf supports and grids to attach without tools or hardware
  • 1.5" (38 mm) slot spacing allows shelves and grids to be adjusted on 1.5" (38 mm) increments
  • Designed for medium-duty applications
  • Two uprights are required for a stand-alone unit
  • Each part number is one individual upright

Corrosion-resistant Metroseal Gray epoxy finish includes Microban® antimicrobial product protections to continuously protect the product and keep it cleaner in between cleanings.

Specifications
  • Depth:
    1⅞"
  • Finish:
    Metroseal epoxy
  • Width:
    1⅛"
  • Color:
    Grey
  • Description:
    Upright
  • Height:
    84½"
  • Material:
    Steel