Orders LinkContactUsLinkComponent
VWR® Calibration Weights
VWR® Calibration Weights
OIML Class F1, VWR-F1-5KG/C, 5 kg
Catalog # 76446-844
Supplier:   VWR International
VWR® Calibration Weights
Catalog # 76446-844
Supplier:  VWR International
VWR® Calibration Weights
VWR® Calibration Weights
OIML Class F1, VWR-F1-5KG/C, 5 kg
Price
$1,421.59
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
VWR® Calibration Weights are crafted from high grade stainless steel and feature a plastic storage case.
  • Option to be supplied with a Statement of Accuracy or an ISO 17025:2017 Certificate of Calibration
  • Cylindrical with knob
  • OIML Class E2, F1, or F2 options available
  • High polish finish
  • Low magnetic permeability
The ISO 17025:2017 Certificate of Calibration provides full traceability of the calibration weight. This certificate provides the date of manufacture, serial number, nominal mass, conventional mass, and uncertainty of measurement.
The Statement of Accuracy provides the date of manufacture, serial number, nominal value of the weight, and accuracy class. Note: the Statement of Accuracy does not provide traceability.
Calibration weights of class E2 and F1 are of solid construction. Calibration weights of Class F2 feature an adjustment cavity.
Certifications: Conforms to OIML-R111 standards. Includes Statement of Accuracy or ISO 17025:2017 Certificate of Calibration.
Delivery information: Includes plastic storage case.
Documents
Specifications
  • Model:
    VWR-F1-5KG/C
  • Description:
    OIML Class F1
  • Certification:
    ISO 17025:2017 Certificate of Calibration
  • Weight:
    5 kg