Orders LinkContactUsLinkComponent
EMS Grid Prep Holders, Electron Microscopy Sciences
Grid prep holder, capacity 81, thickness 6 mm, 65×65 mm
  76439-524
Marketsource
 :  71175-02
EMS Grid Prep Holders, Electron Microscopy Sciences
  76439-524
 :  
Marketsource
EMS Grid Prep Holders, Electron Microscopy Sciences
EMS Grid Prep Holders, Electron Microscopy Sciences
Grid prep holder, capacity 81, thickness 6 mm, 65×65 mm
EMS preparation tray may be use for staining and/or placing an evaporator for the coating of grids.
  • Medical grade aluminum
  • Includes a plastic storage box
These unique trays have a M4 tapped hole in the center which allows for insertion of pin mount. They also have a screw pin in the middle that may be used as a handle for manipulating the trays.
  • Description:
    Grid prep holder, capacity 81, thickness 6 mm
  • Dimensions:
    65×65 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.