Orders LinkContactUsLinkComponent
EMS24 TEM Grid Storage Box, Electron Microscopy Sciences
EMS24 TEM Grid Storage Box, Electron Microscopy Sciences
25 slots, numbered
Catalog # 76439-504
Supplier #:  71159-20
EMS24 TEM Grid Storage Box, Electron Microscopy Sciences
Catalog # 76439-504
EMS24 TEM Grid Storage Box, Electron Microscopy Sciences
EMS24 TEM Grid Storage Box, Electron Microscopy Sciences
25 slots, numbered
Price
$19.45
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The EMS25 is a small capacity grid storage box, designed as a low alternative to larger grid boxes where extra storage capacity is not required. Our grid boxes are intended for use in both routine grid handling and for long term grid storage. Their compact flat shape and low cost make them well suited for the transportation of coated grid products and for the long term storage of catalogued specimens.
The box has a sliding cover, allowing access to five slots at a time. Each diamond shaped hole is capable of storing a 3.05 or 2.3 mm diameter grid. Storage referencing is managed via recessed alpha numeric indexing on the sides of the box.
Specifications
  • Description:
    25 slots, numbered
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)