Orders LinkContactUsLinkComponent
EMS24 TEM Grid Storage Box, Electron Microscopy Sciences
25 slots
  76439-500
 :  71159
EMS24 TEM Grid Storage Box, Electron Microscopy Sciences
  76439-500
EMS24 TEM Grid Storage Box, Electron Microscopy Sciences
EMS24 TEM Grid Storage Box, Electron Microscopy Sciences
25 slots
$19.19
Each
The EMS25 is a small capacity grid storage box, designed as a low alternative to larger grid boxes where extra storage capacity is not required. Our grid boxes are intended for use in both routine grid handling and for long term grid storage. Their compact flat shape and low cost make them well suited for the transportation of coated grid products and for the long term storage of catalogued specimens.
The box has a sliding cover, allowing access to five slots at a time. Each diamond shaped hole is capable of storing a 3.05 or 2.3 mm diameter grid. Storage referencing is managed via recessed alpha numeric indexing on the sides of the box.
  • Description:
    25 slots
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.