Orders LinkContactUsLinkComponent
Universal Solids Standard, Environmental Express®
Universal Soilds Standard, Low Range, 10/pk, Bottle
Catalog # 76436-954
 :  F95005
Universal Solids Standard, Environmental Express®
Catalog # 76436-954
CAS Number:  
Universal Solids Standard, Environmental Express®
Universal Solids Standard, Environmental Express®
Universal Soilds Standard, Low Range, 10/pk, Bottle
CAS Number:  
Price
$175.34
Log in for availability
Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Universal Solids Standard (USS) provides known QC data for all solids procedures.
  • Obtain batch level quality analysis and control for all solids testing at an affordable price
  • Quality guaranteed—NIST-traceability is shown on the Certificate of Analysis and each parameter has an acceptability range of ±3 standard deviations
  • Choose from three ranges to best fit your testing needs
This product complies with the following methods: SM 2540,ASTM D5907
These Environmental Express universal solids standards cover multiple EPA test method standard control requirements including TS, TSS, VS, and VSS. Use is easy—simply add the bottle contents into your graduated cylinder and fill to volume, or use as a Matrix Spike by adding a bottle directly to your sample.
Certifications: Certificate of Analysis with traceability to NIST, SDS, and dilution instructions.
Specifications
Specifications
  • Pack type:
    Bottle
  • Description:
    Universal Soilds Standard, Low Range
  • Size:
    10/pk
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)