SimpleWater™ BOD Nutrient Buffer Solutions, Environmental Express®
Product Details & Documents
No mess - simply pour contents into your container and add water.
- Solutions provide optimal growth media for bacteria in the BOD test
- Borosilicate glass vials with leakproof screw caps
BOD SimpleWater™ Nutrient Buffer Dilution Vials are available in five standard concentrations. BOD dilution water is composed of a variety of nutrients that provide an optimal growth media for bacteria in the BOD test. SimpleWater BOD nutrient buffer solutions are concentrates of the four required nutrient solutions combined in one vial. The contents of the SimpleWater vial are then diluted to the appropriate volume to make up your BOD dilution water.
: Vials are borosilicate glass with lined, leak-proof screw caps. Glass vials are easy-to-rinse with no residue trapping.
Specifications
Specifications
- Description:BOD: SimpleWater™ (Custom Size)
- Size:16 L
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.