Order Entry
Canada
Orders LinkContactUsLinkComponent
SimpleWater™ BOD Nutrient Buffer Solutions, Environmental Express®

SimpleWater™ BOD Nutrient Buffer Solutions, Environmental Express®

SimpleWater™ BOD Nutrient Buffer Solution, 6 L
Catalog # 76436-872
Supplier #:  D4006L
SimpleWater™ BOD Nutrient Buffer Solutions, Environmental Express®
Catalog # 76436-872
CAS Number:  
SimpleWater™ BOD Nutrient Buffer Solutions, Environmental Express®
SimpleWater™ BOD Nutrient Buffer Solutions, Environmental Express®

SimpleWater™ BOD Nutrient Buffer Solution, 6 L

CAS Number:  
Price
$123.38
Log in for availability
Pack of 36 (6l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

No mess - simply pour contents into your container and add water.

  • Solutions provide optimal growth media for bacteria in the BOD test
  • Borosilicate glass vials with leakproof screw caps

BOD SimpleWater™ Nutrient Buffer Dilution Vials are available in five standard concentrations. BOD dilution water is composed of a variety of nutrients that provide an optimal growth media for bacteria in the BOD test. SimpleWater BOD nutrient buffer solutions are concentrates of the four required nutrient solutions combined in one vial. The contents of the SimpleWater vial are then diluted to the appropriate volume to make up your BOD dilution water.

Packaging: Vials are borosilicate glass with lined, leak-proof screw caps. Glass vials are easy-to-rinse with no residue trapping.

Specifications

Specifications

  • Description:
    SimpleWater™ BOD Nutrient Buffer Solution
  • Size:
    6 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.