Ionic Strength Adjustment Buffers, SPEX CertiPrep
Product Details & Documents
Buffers for Ion Chromarography and Ion Selective Electrode Standards
Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary. SPEX CertiPrep offers high quality IC standards available for the analytical laboratory.
Specifications
Specifications
- Pack type:LDPE bottle
- Description:10 M Sodium hydroxide buffer solution
- Size:500 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)