Order Entry
Canada
Orders LinkContactUsLinkComponent
Ionic Strength Adjustment Buffers, SPEX CertiPrep

Ionic Strength Adjustment Buffers, SPEX CertiPrep

5M Sodium nitrate buffer solution, 500 ml, LDPE bottle
Catalog # SXIS-BUF1-500
Supplier:   SPEX CERTIPREP LLC
Supplier #:  IS-BUF1-500
Ionic Strength Adjustment Buffers, SPEX CertiPrep
Catalog # SXIS-BUF1-500
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Ionic Strength Adjustment Buffers, SPEX CertiPrep

5M Sodium nitrate buffer solution, 500 ml, LDPE bottle

CAS Number:  
Price
$307.62
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Buffers for Ion Chromarography and Ion Selective Electrode Standards

Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary. SPEX CertiPrep offers high quality IC standards available for the analytical laboratory.

Specifications

Specifications

  • Pack type:
    LDPE bottle
  • Description:
    5M Sodium nitrate buffer solution
  • Size:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.