Specimen Stages, Electron Microscopy Sciences
Product Details & Documents
EMS Specimen Stages that meets most requirements.
These specimen stages are all easy-change, drop-in style with no scews, and are height adjustable, except for the rotary planetary stage.
Specifications
- Description:Adjusts to accept 6 and 8" wafers comes with 76421-766, an 8" stub holder to accept up to 54¹/₈" pin stubs
- Stage:8" wafer
- For use with:Q300T T plus
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.