Specimen Stages, Electron Microscopy Sciences
Product Details & Documents
EMS Specimen Stages that meets most requirements.
These specimen stages are all easy-change, drop-in style with no scews, and are height adjustable, except for the rotary planetary stage.
Specifications
- Description:Rotate-tilt stage with six stub positions for 15 or 6.5 mm or ¹/₈" pin stubs. Tilts Up to 90° from horizontal
- Stage:Tilt
- For use with:All series
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)