Orders LinkContactUsLinkComponent
Specimen Stages, Electron Microscopy Sciences
Specimen Stages, Electron Microscopy Sciences
9 place coveslip, All series, Stage to accept 9, 20×20 cover slips; The top part of stage lifts off and has a mechanism to lift the cover slips for easy removal
Catalog # 76421-748
Supplier #:  4500-15
Specimen Stages, Electron Microscopy Sciences
Catalog # 76421-748
Specimen Stages, Electron Microscopy Sciences
Specimen Stages, Electron Microscopy Sciences
9 place coveslip, All series, Stage to accept 9, 20×20 cover slips; The top part of stage lifts off and has a mechanism to lift the cover slips for easy removal
Price
$9,771.93
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
EMS Specimen Stages that meets most requirements.
These specimen stages are all easy-change, drop-in style with no scews, and are height adjustable, except for the rotary planetary stage.
Specifications
  • Description:
    Stage to accept 9, 20×20 cover slips; The top part of stage lifts off and has a mechanism to lift the cover slips for easy removal
  • Stage:
    9 place coveslip
  • For use with:
    All series
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.