Orders LinkContactUsLinkComponent
Specimen Stages, Electron Microscopy Sciences
Specimen Stages, Electron Microscopy Sciences
Fiber stage, Q150R plus, Q150T, Stage accepts single fibres or pins up to 1 mm diameter rotating horizontally to achieve all round coating
  76421-738
Supplier #:  4500-10
Specimen Stages, Electron Microscopy Sciences
  76421-738
Specimen Stages, Electron Microscopy Sciences
Specimen Stages, Electron Microscopy Sciences
Fiber stage, Q150R plus, Q150T, Stage accepts single fibres or pins up to 1 mm diameter rotating horizontally to achieve all round coating
$12,806.82
Each
EMS Specimen Stages that meets most requirements.
These specimen stages are all easy-change, drop-in style with no scews, and are height adjustable, except for the rotary planetary stage.
  • Description:
    Stage accepts single fibres or pins up to 1 mm diameter rotating horizontally to achieve all round coating
  • Stage:
    Fiber stage
  • For use with:
    Q150R plus, Q150T
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.