REPLACEMENT GASKET 2NOS F/ELECTRODE CORE
Catalog # 76414-844
Supplier: Azure Biosystems
REPLACEMENT GASKET 2NOS F/ELECTRODE CORE
Catalog # 76414-844
Supplier: Azure Biosystems
Supplier Number: AASP-0021
Frequently Bought Together
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
Water, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
Optical Lens Cleaner, FoxWest Sales
Supplier: Decon Labs
Starch 1% (w/v) in aqueous solution for iodometric titrations mercury-free
Supplier: Ricca Chemical
MBP DNA AWAY™
Supplier: Thermo Scientific
Tryptone to 2xYT Broth, Life Technologies
Supplier: BD
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Masterflex® Hose Clamps, Acetal Copolymer, Avantor®
Supplier: VWR International
COVER GLASS R20-26 RECT PK10
Supplier: Almedic Limited
Specimen Mount Holders, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Picric acid ∼1.2% (w/v) saturated solution in water
Supplier: Ricca Chemical
N,N-Dimethylformamide ≥99.8% (by GC), GR ACS, Supelco®
Supplier: MilliporeSigma
Ammonium dihydrogen phosphate ≥99.0% (by acidimetry), EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
PCR STRIP MAGSTAND
Supplier: Zymo Research