Order Entry
Canada
Orders LinkContactUsLinkComponent
 

VWR® Certified Reference Materials, Oxide

2 kg, Gold, >100 ppm, Au, Oxide, Jar
Catalog # 76411-358
 :  21001491
VWR® Certified Reference Materials, Oxide
Catalog # 76411-358
CAS Number:  
VWR® Certified Reference Materials, Oxide
VWR® Certified Reference Materials, Oxide

2 kg, Gold, >100 ppm, Au, Oxide, Jar

CAS Number:  
Price
504,89 $
Log in for availability
Each (2,000g) Case of 4 (2,000,g)
Product Details & Documents
Specifications

Specifications

  • Concentration:
    >100 ppm
  • Matrix:
    Oxide
  • Pack type:
    Jar
  • Size:
    2 kg
  • Analyte:
    Au
  • Element:
    Gold

Specifications