Order Entry
Canada
ContactUsLinkComponent
Tryptic Soy Agar (TSA), SterEM*, irradiated, USP, triple bagged, Dimension: 15x100mm plate, Volunme: 26ml fill, for use as a general growth medium for the detection and enumeration of microorganisms
SterEM™ Tryptic Soy Agar (TSA), USP, Irradiated
Catalog # CA76200-286
CAS Number:  
undefined
SterEM™ Tryptic Soy Agar (TSA), USP, Irradiated
Catalog # CA76200-286
Supplier Number:  W570
CAS Number:  

Specifications

  • Pack type:
    10/pack
  • Description:
    SterEM™ Tryptic soy agar (TSA), irradiated, USP, triple bagged
  • Dimensions:
    1.5×10 cm
  • Cat. no.:
    CA76200-286
  • Sterility:
    Promotes a higher sterility assurance level of 10⁻⁶
  • Application:
    Compounding pharmacy, pharmaceutical manufacturing
  • Volume:
    26 ml

Specifications

About this item

Sterile plates for environmental monitoring.

  • New moisture control packaging format prevents moisture buildup and ensures safe and sterile transfer
  • Optically clear polystyrene for distortion free viewing
  • Heavy-Duty - Reduces breakage during shipping
  • Polystyrene petri plates feature raised rings for stacking ability and prevents sliding
  • Sterile by irradiation - Validated for Sterility Assurance Level (SAL) of 10-5
  • Triple bagged, two nylon inner bags and one outer Mylar(R) bag that is heat sealed

Irradiated Tryptic Soy Agar (TSA), USP is recommended for use as a general growth medium for the detection and enumeration of microorganisms. The plates are triple bagged and sterilized by irradiation to promote a higher sterility assurance level.

Certifications: Certified to ISO 13485 - Manufactured in a licensed cGMP facility with the quality management system certified to ISO 13485.
Frequently Bought Together