AccuStart™ Long Range SuperMix, Quantabio
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Superior sensitivity and multiplexing for DNA amplification of long targets.
- Amplify +24 kb gDNA and +40 kb lambda DNA
- 4X concentration enables superior sensitivity with low inputs (100 pg)
- Multiplex up to 6 targets with each target up to 6 kb in length
- Stabilized, single-tube SuperMix minimizes pipetting errors and hands-on-time
- Reliable lot-to-lot reproducibility ensures consistent results
- Manufactured in state of the art ISO 13485 facility
The AccuStart Long Range SuperMix is a 4X, ready-to-use solution that contains all the components for long range target amplifications, including a blend of two hot-start thermostable DNA polymerases (one with strong proof-reading activity) and an optimized buffer to ensure high efficiency, sensitivity and specificity.
The SuperMix enables routine and easy amplification of up to 24 kb target from gDNA with high accuracy (>10-fold better fidelity than Taq) and accommodates targets with broad GC-content (no separate GC buffer needed). This product is also capable of amplifying multiple targets simultaneously and is suitable for end point PCR, template prep for sanger sequencing, NGS, cloning and HLA typing. Features improved yields across a range of DNA inputs and target sizes with inputs as low as 100 pg, strong yields at 24 kb length as well as clean and visible PCR bands.
This can multiplex 6 targets up to 6 kb each, further speeding up experiments, reducing costs and allowing for more data to be derived per run, reducing costs and increasing the speed of experiments while generating more data per run.
Documents
Specifications
Specifications
- Description:AccuStart™ Long Range SuperMix
- Size:25 reactions (12.5 µl)
- Cat. No.:CA76359-896
- Storage conditions:−25...−15 °C