Order Entry
Canada
Orders LinkContactUsLinkComponent
TitroLine® 5000 Titrators, SI Analytics

TitroLine® 5000 Titrators, SI Analytics

TitroLine® 5000 M1 with 50 ml dosing unit and magnetic stirrer (TM 235)
Catalog # 76403-686
Supplier:   YSI
Supplier #:  285230150
TitroLine® 5000 Titrators, SI Analytics
Catalog # 76403-686
Supplier:  YSI
TitroLine® 5000 Titrators, SI Analytics

TitroLine® 5000 M1 with 50 ml dosing unit and magnetic stirrer (TM 235)

Price
$11,882.33
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

The TitroLine® 5000, successor of the TitroLine® easy, is a very easy to use automatic titrator for a variety of applications, including water/wastewater applications, environmental and food analysis.

  • High resolution pH/mV measurement input for pH, ORP, silver and further mV electrodes
  • PT1000 and NTC 30 temperature measurement input for automatic temperature compensation
  • Pre-installed standard methods for FOS/TAC, alkalinity, total acidity in drinks, chloride
  • Linear and dynamic titration to equivalence point
  • Titration to pH and mV endpoints
  • Same manual titration and dosing function as the Titronic® 300 piston burette

Delivery information: Basic (M1) units supplied with pre-assembled 20 or 50 ml dosing unit, manual controller TZ 3880, titration clamp, stand rod, magnetic stirrer TM 50 and power supply. M2 units also include a pH electrode and buffer set. The M3 unit includes an Ag electrode instead of a pH electrode.

Specifications
  • Description:
    TitroLine® 5000 M1 with 50 ml dosing unit and magnetic stirrer (TM 235)
  • Connections:
    1× USB-A, 1× USB-B, 2× RS 232C
  • Dosing volume:
    20/50 ml
  • Compliance:
    ISO 8655, part 6; EMC: 2004/108/EG; safety EG Directive 2006/95
  • Dosing accuracy:
    0.15% (systematic error); 0.05% (random error)
  • Volume display:
    0000.000 - 9999.999 ml
  • Measuring input electrode:
    pH/mV input with electrode socket according to DIN 19 262
  • Power supply:
    100 - 240 V, 50/60 Hz
  • Dosing speed:
    Max. 100 ml/min (with 50 ml unit)
  • Filling time:
    30 s - 999 s (time according to the cylinder volume)
  • Weight:
    2 kg (4.4 lbs.)
  • Cat. No.:
    76403-686
  • Stirrer connection:
    Connection for stirrer TM 50 (included)
  • Measuring input PT1000:
    Socket 2×4 mm (PT1000/NTC 30)
  • Keypad:
    Optional USB keyboard (TZ 3835) can be connected
  • Model:
    TitroLine® 5000
  • W×D×H:
    13.5×20.5×31.0 cm
  • mV accuracy:
    −1900 to 1900…1.0 ±1
  • Display resolution:
    0.005 - 0.025 ml (depending on dosing unit)
  • Burette resolution:
    8000 steps
  • Display:
    Graphical TFT
  • Operating temperature:
    10...40 °C (50...104 °F)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.